2-Methyl-2-(4-vinylphenyl)propanoic acid ≥98%
Dodavatel: Apollo Scientific
OR370067-100MG
OR370067-250MG
APOSOR370067-100MGEA
14190
CZK
APOSOR370067-100MG
APOSOR370067-250MG
2-Methyl-2-(4-vinylphenyl)propanoic acid ≥98%
2-Methyl-2-(4-vinylphenyl)propanoic acid
Vzorec:
C₁₂H₁₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD30377216 CAS číslo: 1256584-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...