2-Bromoimidazo[2,1-b]thiazole ≥98%
Dodavatel: Apollo Scientific
OR370065-100MG
OR370065-250MG
APOSOR370065-100MGEA
4040
CZK
APOSOR370065-100MG
APOSOR370065-250MG
2-Bromoimidazo[2,1-b]thiazole ≥98%
2-Bromoimidazo[2,1-b]thiazole
Vzorec:
C₅H₃BrN₂S MW: 203,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11036022 CAS číslo: 944581-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...