Tisk…

(1R,2R)-2-((R)-1-Phenylethylamino)cyclohexanol ≥98%

Dodavatel: Apollo Scientific

OR370064-100MG OR370064-250MG
APOSOR370064-100MGEA 2690 CZK
APOSOR370064-100MG APOSOR370064-250MG
(1R,2R)-2-((R)-1-Phenylethylamino)cyclohexanol ≥98%
(1R,2R)-2-((R)-1-Phenylethylamino)cyclohexanol
Vzorec: C₁₄H₂₁NO
Storage Temperature: Ambient
MDL Number: MFCD18642889
CAS číslo: 98462-58-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR