Tisk…

2-(3-Amino-5-methoxyphenyl)ethanol ≥98%

Dodavatel: Apollo Scientific

OR370059-100MG OR370059-1G
APOSOR370059-100MGEA 10320 CZK
APOSOR370059-100MG APOSOR370059-1G
2-(3-Amino-5-methoxyphenyl)ethanol ≥98%
2-(3-Amino-5-methoxyphenyl)ethanol
Vzorec: C₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD18642813
CAS číslo: 354512-41-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR