Tisk…

3-Hydroxy-3,3a,4,6a-tetrahydro-2H-cyclopenta[b]furan-2-one ≥98%

Dodavatel: Apollo Scientific

OR370046-1G OR370046-250MG
APOSOR370046-1GEA 32500 CZK
APOSOR370046-1G APOSOR370046-250MG
3-Hydroxy-3,3a,4,6a-tetrahydro-2H-cyclopenta[b]furan-2-one ≥98%
3-Hydroxy-3,3a,4,6a-tetrahydro-2H-cyclopenta[b]furan-2-one
Vzorec: C₇H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD02094031
CAS číslo: 189574-94-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR