2-(Bromomethyl)-5-(tert-butyl)oxazole ≥98%
Dodavatel: Apollo Scientific
OR370036-100MG
OR370036-250MG
OR370036-500MG
APOSOR370036-100MGEA
14490
CZK
APOSOR370036-100MG
APOSOR370036-250MG
APOSOR370036-500MG
2-(Bromomethyl)-5-(tert-butyl)oxazole ≥98%
2-(Bromomethyl)-5-(tert-butyl)oxazole
Vzorec:
C₈H₁₂BrNO MW: 218.091 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20231570 CAS číslo: 1334492-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...