Tisk…

4-Bromo-1-methyl-2-phenyl-1H-imidazole ≥98%

Dodavatel: Apollo Scientific

OR370029-5G OR370029-100MG OR370029-250MG OR370029-1G
APOSOR370029-5GEA 54550 CZK
APOSOR370029-5G APOSOR370029-100MG APOSOR370029-250MG APOSOR370029-1G
4-Bromo-1-methyl-2-phenyl-1H-imidazole ≥98%
4-Bromo-1-methyl-2-phenyl-1H-imidazole
Vzorec: C₁₀H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD13811097
CAS číslo: 86119-59-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR