Tisk…

1-Benzyl-1H-pyrrolo[2,3-b]pyridin-4-amine ≥98%

Dodavatel: Apollo Scientific

OR370026-100MG OR370026-1G
APOSOR370026-100MGEA 11010 CZK
APOSOR370026-100MG APOSOR370026-1G
1-Benzyl-1H-pyrrolo[2,3-b]pyridin-4-amine ≥98%
1-Benzyl-1H-pyrrolo[2,3-b]pyridin-4-amine
Vzorec: C₁₄H₁₃N₃
Storage Temperature: Ambient
MDL Number: MFCD15529357
CAS číslo: 152955-55-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR