Tisk…

p-Hydroxyphenyl propanoate ≥98%

Dodavatel: Apollo Scientific

OR370020-10G OR370020-5G
APOSOR370020-10GEA 29890 CZK
APOSOR370020-10G APOSOR370020-5G
p-Hydroxyphenyl propanoate ≥98%
p-Hydroxyphenyl propanoate
Vzorec: C₉H₁₀O₃
MW: 166,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042386
CAS číslo: 3233-34-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR