Tisk…

3-(Phenylmethyl)-3-azabicyclo[3.1.0]hexane-1-carbonitrile ≥98%

Dodavatel: Apollo Scientific

OR370018-100MG OR370018-250MG
APOSOR370018-100MGEA 11970 CZK
APOSOR370018-100MG APOSOR370018-250MG
3-(Phenylmethyl)-3-azabicyclo[3.1.0]hexane-1-carbonitrile ≥98%
3-(Phenylmethyl)-3-azabicyclo[3.1.0]hexane-1-carbonitrile
Vzorec: C₁₃H₁₄N₂
Storage Temperature: Ambient
MDL Number: MFCD11042334
CAS číslo: 56062-59-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR