1-(Imidazo[1,2-a]pyridin-6-yl)ethanone ≥98%
Dodavatel: Apollo Scientific
OR370013-100MG
OR370013-1G
OR370013-250MG
APOSOR370013-100MGEA
9560
CZK
APOSOR370013-100MG
APOSOR370013-1G
APOSOR370013-250MG
1-(Imidazo[1,2-a]pyridin-6-yl)ethanone ≥98%
1-(Imidazo[1,2-a]pyridin-6-yl)ethanone
Vzorec:
C₉H₈N₂O Storage Temperature: Ambient |
MDL Number:
MFCD10696290 CAS číslo: 944905-12-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...