Tisk…

4-Bromophthalazin-1(2H)-one ≥98%

Dodavatel: Apollo Scientific

OR370012-100MG OR370012-1G OR370012-250MG
APOSOR370012-100MGEA 612 CZK
APOSOR370012-100MG APOSOR370012-1G APOSOR370012-250MG
4-Bromophthalazin-1(2H)-one ≥98%
Vzorec: C₈H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD11101043
CAS číslo: 19064-73-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR