Tisk…

Ethyl 2-bromothiophene-3-carboxylate ≥98%

Dodavatel: Apollo Scientific

OR370011-25G OR370011-5G
APOSOR370011-25GEA 18250 CZK
APOSOR370011-25G APOSOR370011-5G
Ethyl 2-bromothiophene-3-carboxylate ≥98%
Ethyl 2-bromothiophene-3-carboxylate
Vzorec: C₇H₇BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD09965551
CAS číslo: 632325-50-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR