Tisk…

4-Acridinecarboxylic acid ≥98%

Dodavatel: Apollo Scientific

Synonyms: Acridine-4-carboxylic acid

OR370004-100MG OR370004-1G OR370004-250MG
APOSOR370004-100MGEA 2240 CZK
APOSOR370004-100MG APOSOR370004-1G APOSOR370004-250MG
4-Acridinecarboxylic acid ≥98%
4-Acridinecarboxylic acid
Vzorec: C₁₄H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD21340176
CAS číslo: 31327-97-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR