Tisk…

2-Amino-3-phenylquinoline

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Aminoquinolin-3-yl)benzene, 3-Phenylquinolin-2-amine

OR3652-1G OR3652-500MG
APOSOR3652-1GEA 10580 CZK
APOSOR3652-1G APOSOR3652-500MG
2-Amino-3-phenylquinoline
2-Amino-3-phenylquinoline
Vzorec: C₁₅H₁₂N₂
MW: 220.269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272522
CAS číslo: 36926-84-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR