Tisk…

(2R,3R)-2-Phenylpyrrolidine-3-carboxylic acid ≥98%

Dodavatel: Apollo Scientific

OR361744-100MG OR361744-250MG
APOSOR361744-100MGEA 18620 CZK
APOSOR361744-100MG APOSOR361744-250MG
(2R,3R)-2-Phenylpyrrolidine-3-carboxylic acid ≥98%
(2R,3R)-2-Phenylpyrrolidine-3-carboxylic acid
Vzorec: C₁₁H₁₃NO₂
MW: 191,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374329
CAS číslo: 1241684-18-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR