Tisk…

Ethyl (1S,2R)-2-aminocyclopentane-1-carboxylate hydrochloride ≥98%

Dodavatel: Apollo Scientific

OR361741-1G OR361741-250MG OR361741-5G
APOSOR361741-1GEA 8340 CZK
APOSOR361741-1G APOSOR361741-250MG APOSOR361741-5G
Ethyl (1S,2R)-2-aminocyclopentane-1-carboxylate hydrochloride ≥98%
Ethyl (1S,2R)-2-aminocyclopentane-1-carboxylate hydrochloride
Vzorec: C₈H₁₅NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD28656858
CAS číslo: 945935-60-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR