Tisk…

3-(5-Bromo-2-hydroxyphenyl)propanoic acid ≥98%

Dodavatel: Apollo Scientific

OR361737-100MG OR361737-250MG
APOSOR361737-100MGEA 26420 CZK
APOSOR361737-100MG APOSOR361737-250MG
3-(5-Bromo-2-hydroxyphenyl)propanoic acid ≥98%
3-(5-Bromo-2-hydroxyphenyl)propanoic acid
Vzorec: C₉H₉BrO₃
MW: 245,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09705973
CAS číslo: 749878-59-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR