Tisk…

3-Phenyl-1-propyl-1H-pyrazol-5-amine ≥98%

Dodavatel: Apollo Scientific

OR361730-100MG OR361730-1G OR361730-250MG
APOSOR361730-100MGEA 6750 CZK
APOSOR361730-100MG APOSOR361730-1G APOSOR361730-250MG
3-Phenyl-1-propyl-1H-pyrazol-5-amine ≥98%
3-Phenyl-1-propyl-1H-pyrazol-5-amine
Vzorec: C₁₂H₁₅N₃
MW: 201,268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14609384
CAS číslo: 3524-48-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR