Tisk…

N-Allylcyanamide ≥98%

Dodavatel: Apollo Scientific

OR361719-100MG OR361719-250MG
APOSOR361719-100MGEA 15180 CZK
APOSOR361719-100MG APOSOR361719-250MG
N-Allylcyanamide ≥98%
N-Allylcyanamide
Vzorec: C₄H₆N₂
Storage Temperature: Ambient
MDL Number: MFCD16767888
CAS číslo: 36333-01-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR