Quinazolin-6-ol ≥98%
Dodavatel: Apollo Scientific
OR361718-100MG
OR361718-250MG
APOSOR361718-100MGEA
33400
CZK
APOSOR361718-100MG
APOSOR361718-250MG
Quinazolin-6-ol ≥98%
Quinazolin-6-ol
Vzorec:
C₈H₆N₂O MW: 146.146 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00456081 CAS číslo: 7556-93-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...