Tisk…

2-(4-Methylpiperazino)pyrimidine-5-boronic acid

Dodavatel: Apollo Scientific

Synonyms: (2-(4-Methylpiperazin-1-yl)pyrimidin-5-yl)boronic acid

OR361463-1G OR361463-250MG
APOSOR361463-1GEA 8770 CZK
APOSOR361463-1G APOSOR361463-250MG
2-(4-Methylpiperazino)pyrimidine-5-boronic acid
2-(4-Methylpiperazino)pyrimidine-5-boronic acid
Vzorec: C₉H₁₅BN₄O₂
MW: 222,052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375170
CAS číslo: 1256355-29-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR