Tisk…

4-(1-Cyanocyclopropyl)phenylboronic acid

Dodavatel: Apollo Scientific

Synonyms: (4-(1-Cyanocyclopropyl)phenyl)boronic acid

OR361008-1G OR361008-250MG
APOSOR361008-1GEA 7080 CZK
APOSOR361008-1G APOSOR361008-250MG
4-(1-Cyanocyclopropyl)phenylboronic acid
4-(1-Cyanocyclopropyl)phenylboronic acid
Vzorec: C₁₀H₁₀BNO₂
Storage Temperature: Ambient
MDL Number: MFCD12546546
CAS číslo: 1217501-00-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR