Tisk…

2-Chloro-6-methylpyridine-4-boronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Chloro-6-methylpyridin-4-yl)boronic acid

OR360974-1G OR360974-5G
APOSOR360974-1GEA 11580 CZK
APOSOR360974-1G APOSOR360974-5G
2-Chloro-6-methylpyridine-4-boronic acid 98%
2-Chloro-6-methylpyridine-4-boronic acid
Vzorec: C₆H₇BClNO₂
MW: 171,389 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13188704
CAS číslo: 1320397-15-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR