1-tert-Butylpyrazole-4-boronic acid, pinacol ester 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-t-Butylpyrazole-4-boronic acid, pinacol ester, 1-(tert-Butyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
OR360855-250MG
OR360855-1G
APOSOR360855-250MGEA
2440
CZK
APOSOR360855-250MG
APOSOR360855-1G
1-tert-Butylpyrazole-4-boronic acid, pinacol ester 97%
1-tert-Butylpyrazole-4-boronic acid, pinacol ester
Vzorec:
C₁₃H₂₃BN₂O₂ MW: 250,145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16660302 CAS číslo: 1256359-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...