Tisk…

4-Isobutyrylphenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Isobutyrylphenyl)boronic acid

OR360548-1G OR360548-5G
APOSOR360548-1GEA 6450 CZK
APOSOR360548-1G APOSOR360548-5G
4-Isobutyrylphenylboronic acid 97%
4-Isobutyrylphenylboronic acid
Vzorec: C₁₀H₁₃BO₃
MW: 192.019 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12022352
CAS číslo: 186498-27-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR