Tisk…

2-Methyl-4-cyanophenylboronic acid

Dodavatel: Apollo Scientific

Synonyms: 4-Cyano-2-methylphenylboronic acid, (4-Cyano-2-methylphenyl)boronic acid

OR360237-5G
APOSOR360237-5GEA 4600 CZK
APOSOR360237-5G
2-Methyl-4-cyanophenylboronic acid
2-Methyl-4-cyanophenylboronic acid
Vzorec: C₈H₈BNO₂
MW: 160,966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750473
CAS číslo: 313546-18-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR