6-Hydroxy-3-pyridineboronic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR360140-1G
OR360140-250MG
APOSOR360140-1GEA
1120
CZK
APOSOR360140-1G
APOSOR360140-250MG
6-Hydroxy-3-pyridineboronic acid
6-Hydroxy-3-pyridineboronic acid
Vzorec:
C₅H₆BNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09907979 CAS číslo: 903899-13-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...