2-(2-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane 95+%
Dodavatel: Apollo Scientific
Synonyms:
2-Chloro-3-methoxyphenylboronic acid pinacol ester
OR360118-1G
APOSOR360118-1GEA
1660
CZK
APOSOR360118-1G
2-(2-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane 95+%
2-(2-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane
Vzorec:
C₁₃H₁₈BClO₃ MW: 268,544 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07782041 CAS číslo: 1151564-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...