Tisk…

2-Cyanopyridine-3-boronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Cyanopyridin-3-yl)boronic acid

OR360054-1G OR360054-250MG
APOSOR360054-1GEA 13790 CZK
APOSOR360054-1G APOSOR360054-250MG
2-Cyanopyridine-3-boronic acid
2-Cyanopyridine-3-boronic acid
Vzorec: C₆H₅BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08458475
CAS číslo: 874290-88-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR