Tisk…

Benzocyclobutene-4-boronic acid

Dodavatel: Apollo Scientific

OR360030-1G OR360030-250MG
APOSOR360030-1GEA 2820 CZK
APOSOR360030-1G APOSOR360030-250MG
Benzocyclobutene-4-boronic acid
Benzocyclobutene-4-boronic acid
Vzorec: C₈H₇BO₂
Storage Temperature: Ambient
MDL Number: MFCD09752664
CAS číslo: 195730-31-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR