Tisk…

Methyl 4-amino-5-(ethylsulphonyl)-2-methoxybenzoate

Dodavatel: Apollo Scientific
Varování

OR3572-25G OR3572-5G
APOSOR3572-25GEA 1990 CZK
APOSOR3572-25G APOSOR3572-5G
Methyl 4-amino-5-(ethylsulphonyl)-2-methoxybenzoate
Methyl 4-amino-5-(ethylsulphonyl)-2-methoxybenzoate
Vzorec: C₁₁H₁₅NO₅S
Storage Temperature: Ambient
MDL Number: MFCD07787259
CAS číslo: 80036-89-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR