Tisk…

7-Hydroxy-3-(4'-methoxyphenyl)coumarin 98%

Dodavatel: Apollo Scientific
Varování

OR351061-1G OR351061-5G OR351061-250MG
APOSOR351061-1GEA 5630 CZK
APOSOR351061-1G APOSOR351061-5G APOSOR351061-250MG
7-Hydroxy-3-(4'-methoxyphenyl)coumarin 98%
7-Hydroxy-3-(4'-methoxyphenyl)coumarin
Vzorec: C₁₆H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD02093155
CAS číslo: 66267-82-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR