Tisk…

1-(Bromomethyl)-2-methoxybenzene, Technical Grade

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Methoxybenzyl bromide

OR350580-1G OR350580-5G OR350580-250MG
APOSOR350580-1GEA 8900 CZK
APOSOR350580-1G APOSOR350580-5G APOSOR350580-250MG
1-(Bromomethyl)-2-methoxybenzene, Technical Grade
1-(Bromomethyl)-2-methoxybenzene
Vzorec: C₈H₉BrO
MW: 201.06 g/mol
Bod varu: 118 °C (18 mm Hg)
Bod tání: 47…50 °C
Storage Temperature: Freezer
MDL Number: MFCD09745128
CAS číslo: 52289-93-7
UN: 1759
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR