Tisk…

Methyl 1-amino-1H-pyrrole-2-carboxylate

Dodavatel: Apollo Scientific

OR350273-1G OR350273-250MG
APOSOR350273-1GEA 8040 CZK
APOSOR350273-1G APOSOR350273-250MG
Methyl 1-amino-1H-pyrrole-2-carboxylate
Methyl 1-amino-1H-pyrrole-2-carboxylate
Vzorec: C₆H₈N₂O₂
MW: 140,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226170
CAS číslo: 122181-85-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR