Methyl trans-cinnamate 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl (E)-cinnamate, trans-Cinnamic acid methyl ester
OR350234-100G
OR350234-500G
APOSOR350234-100GEA
744
CZK
APOSOR350234-100G
APOSOR350234-500G
Methyl trans-cinnamate 95%
Methyl trans-cinnamate
Vzorec:
C₁₀H₁₀O₂ MW: 162,19 g/mol Bod varu: 262 °C (760 mm Hg) Bod tání: 34 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008458 CAS číslo: 1754-62-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...