Tisk…

4-Formyl-2-methoxyphenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Formyl-2-methoxyphenyl)boronic acid

OR350182-25G OR350182-5G
APOSOR350182-25GEA 23350 CZK
APOSOR350182-25G APOSOR350182-5G
4-Formyl-2-methoxyphenylboronic acid
4-Formyl-2-methoxyphenylboronic acid
Vzorec: C₈H₉BO₄
MW: 179,966 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11856026
CAS číslo: 1028479-47-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR