1,2-Dihydro-3H-1,2,4-triazol-3-one 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1H-1,2,4-Triazol-3(2H)-one
OR350126-100G
OR350126-25G
OR350126-500G
APOSOR350126-100GEA
2070
CZK
APOSOR350126-100G
APOSOR350126-25G
APOSOR350126-500G
1,2-Dihydro-3H-1,2,4-triazol-3-one 98%
1,2-Dihydro-3H-1,2,4-triazol-3-one
Vzorec:
C₂H₃N₃O MW: 85,065 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01311031 CAS číslo: 930-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...