Tisk…

1-Benzothiophene-5-carbonitrile

Dodavatel: Apollo Scientific

OR350057-1G OR350057-250MG
APOSOR350057-1GEA 4700 CZK
APOSOR350057-1G APOSOR350057-250MG
1-Benzothiophene-5-carbonitrile
1-Benzothiophene-5-carbonitrile
Vzorec: C₉H₅NS
MW: 159,208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664404
CAS číslo: 2060-63-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR