2-(4-Bromopyridin-2-yl)propan-2-ol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(4-Bromo-2-pyridinyl)-2-propanol
OR350034-250MG
OR350034-1G
OR350034-5G
APOSOR350034-250MGEA
1910
CZK
APOSOR350034-250MG
APOSOR350034-1G
APOSOR350034-5G
2-(4-Bromopyridin-2-yl)propan-2-ol
2-(4-Bromopyridin-2-yl)propan-2-ol
Vzorec:
C₈H₁₀BrNO MW: 216,075 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13190619 CAS číslo: 477252-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...