2,2-Dichloro-2-phenylacetonitrile 97%
Dodavatel: Apollo Scientific
OR3472-1G
OR3472-5G
APOSOR3472-1GEA
8070
CZK
APOSOR3472-1G
APOSOR3472-5G
2,2-Dichloro-2-phenylacetonitrile 97%
2,2-Dichloro-2-phenylacetonitrile
Vzorec:
C₈H₅Cl₂N MW: 186.038 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09702429 CAS číslo: 40626-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...