2,3-Dihydrobenzofuran-5-carboxamide 95+%
Dodavatel: Apollo Scientific
OR346694-1G
OR346694-5G
APOSOR346694-1GEA
4770
CZK
APOSOR346694-1G
APOSOR346694-5G
2,3-Dihydrobenzofuran-5-carboxamide 95+%
2,3-Dihydrobenzofuran-5-carboxamide
Vzorec:
C₉H₉NO₂ MW: 163,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00114406 CAS číslo: 262847-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...