Tisk…

(Benzofuran-2-yl)acetonitrile 95+%

Dodavatel: Apollo Scientific

OR346625-1G OR346625-500MG
APOSOR346625-1GEA 24970 CZK
APOSOR346625-1G APOSOR346625-500MG
(Benzofuran-2-yl)acetonitrile 95+%
(Benzofuran-2-yl)acetonitrile
Vzorec: C₁₀H₇NO
MW: 157,169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052739
CAS číslo: 95-28-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR