Tisk…

2-Chloro-5-methoxybenzenesulphonyl chloride 95+%

Dodavatel: Apollo Scientific

Synonyms: 2-Chloro-5-methoxybenzene-1-sulphonyl chloride

OR346567-1G OR346567-5G
APOSOR346567-1GEA 5460 CZK
APOSOR346567-1G APOSOR346567-5G
2-Chloro-5-methoxybenzenesulphonyl chloride 95+%
2-Chloro-5-methoxybenzenesulphonyl chloride
Vzorec: C₇H₆Cl₂O₃S
MW: 241,092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026184
CAS číslo: 201935-41-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR