1-p-Tolyl-1H-pyrazole 95+%
Dodavatel: Apollo Scientific
Synonyms:
1-(p-Tolyl)-1H-pyrazole
OR346533-1G
OR346533-5G
APOSOR346533-1GEA
5130
CZK
APOSOR346533-1G
APOSOR346533-5G
1-p-Tolyl-1H-pyrazole 95+%
1-p-Tolyl-1H-pyrazole
Vzorec:
C₁₀H₁₀N₂ MW: 158.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00748760 CAS číslo: 20518-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...