Tisk…

2-Iodo-quinoline-3-carboxaldehyde 95+%

Dodavatel: Apollo Scientific

OR346531-1G OR346531-250MG
APOSOR346531-1GEA 17760 CZK
APOSOR346531-1G APOSOR346531-250MG
2-Iodo-quinoline-3-carboxaldehyde 95+%
2-Iodo-quinoline-3-carboxaldehyde
Vzorec: C₁₀H₆INO
Storage Temperature: Ambient
MDL Number: MFCD18448256
CAS číslo: 80231-40-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR