5-tert-Butyl-2-methyl-2H-pyrazole-3-carbonitrile 95+%
Dodavatel: Apollo Scientific
OR346458-1G
OR346458-5G
APOSOR346458-1GEA
4770
CZK
APOSOR346458-1G
APOSOR346458-5G
5-tert-Butyl-2-methyl-2H-pyrazole-3-carbonitrile 95+%
5-tert-Butyl-2-methyl-2H-pyrazole-3-carbonitrile
Vzorec:
C₉H₁₃N₃ MW: 163,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21362501 CAS číslo: 1196104-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...