2-Chloro-3-phenylpropionitrile 95+%
Dodavatel: Apollo Scientific
OR346453-1G
OR346453-5G
APOSOR346453-1GEA
6190
CZK
APOSOR346453-1G
APOSOR346453-5G
2-Chloro-3-phenylpropionitrile 95+%
2-Chloro-3-phenylpropionitrile
Vzorec:
C₉H₈ClN MW: 165,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00995426 CAS číslo: 17849-62-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...