4-(2,2,2-Trichloroacetyl)-1H-pyrrole-2-carbonitrile 95+%
Dodavatel: Apollo Scientific
OR346430-1G
OR346430-5G
APOSOR346430-1GEA
6190
CZK
APOSOR346430-1G
APOSOR346430-5G
4-(2,2,2-Trichloroacetyl)-1H-pyrrole-2-carbonitrile 95+%
4-(2,2,2-Trichloroacetyl)-1H-pyrrole-2-carbonitrile
Vzorec:
C₇H₃Cl₃N₂O MW: 237,471 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19981427 CAS číslo: 66832-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...