Tisk…

1-(4-Bromophenyl)-1H-pyrazole-4-carboxaldehyde 95+%

Dodavatel: Apollo Scientific

OR346398-1G OR346398-500MG
APOSOR346398-1GEA 10910 CZK
APOSOR346398-1G APOSOR346398-500MG
1-(4-Bromophenyl)-1H-pyrazole-4-carboxaldehyde 95+%
1-(4-Bromophenyl)-1H-pyrazole-4-carboxaldehyde
Vzorec: C₁₀H₇BrN₂O
MW: 251,079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426979
CAS číslo: 294877-41-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR